Alkaline Phosphatase, calf intestine, EIA Grade - FAQs
View additional product information for Alkaline Phosphatase, calf intestine, EIA Grade - FAQs (J61037.LB0, J61037.MCR)
11 product FAQs found
What are the applications of BioAcryl-P (30%, 37.5:1) (Cat. No. J61505.K2, J61505-K2)?
BioAcryl-P (30%, 37.5:1) is suitable for conjugation to antibodies and other proteins for ELISA, Western blotting, and other histochemical procedures. Alkaline phosphatase may be used to dephosphorylate the 5' ends of DNA or RNA. It can also be used to dephosphorylate proteins.
What is the specific activity of BioAcryl-P (30%, 37.5:1) (Cat. No. J61505.K2, J61505-K2)?
The specific activity of BioAcryl-P (30%, 37.5:1) is ≥3,000 units/mg protein (DEA units).
What is the enzyme affinity (Km) of BioAcryl-P (30%, 37.5:1) (Cat. No. J61505.K2, J61505-K2)?
The Km is 1.5 x 10-3 M (p-Nitrophenyl phosphate); 19 x 10-3 M (phosphoenolypyruvate).
What is the extinction coefficient of BioAcryl-P (30%, 37.5:1) (Cat. No. J61505.K2, J61505-K2)?
The extinction coefficient (278nm) of BioAcryl-P (30%, 37.5:1) is 7.6-10.5.
What is the molecular mass of BioAcryl-P (30%, 37.5:1) (Cat. No. J61505.K2, J61505-K2)?
The molecular mass of BioAcryl-P (30%, 37.5:1) is ~ 140 kDa.
What is the enzyme commission number (EC) of BioAcryl-P (30%, 37.5:1) (Cat. No. J61505.K2, J61505-K2)?
The EC for BioAcryl-P (30%, 37.5:1) is 3.1.3.1.
What is the definition of one unit of Alkaline Phosphatase, calf intestine, EIA Grade (Cat. Nos. J61037.xx)?
One unit is defined as the amount of protein that hydrolyzes 1 µmole of 4-nitrophenyl phosphate per minute at pH 9.8 and 37°C. One glycine unit is equal to ~3 DEA units.
What is the optimum pH for Alkaline Phosphatase, calf intestine, EIA Grade (Cat. Nos. J61037.xx)?
Optimum pH varies depending on the substrate, substrate concentration, and the ionic concentration. The enzyme is most stable at a pH range of 7.5-9.5. The optimum pH range for catalytic activity is 8-10. The enzyme has optimal activity using Diethanolamine (DEA) as a substrate at a pH of ~9.8.
How can I inhibit Alkaline Phosphatase enzyme activity for Alkaline Phosphatase, calf intestine, EIA Grade (Cat. Nos. J61037.xx)?
The enzyme activity can be inhibited by addition of chelating agents or inorganic phosphates.
What are the concentrations of the components in Alkaline Phosphatase, calf intestine, EIA Grade (Cat. Nos. J61037.xx)?
Alkaline Phosphatase, calf intestine, EIA Grade is supplied as a 20 mg/mL protein solution in 50% glycerol, containing 5 mM Tris, 5 mM MgCl2 and 0.12 mM ZnCl2.
What storage conditions should I use for Alkaline Phosphatase, calf intestine, EIA Grade (Cat. Nos. J61037.xx)?
Alkaline Phosphatase, calf intestine, EIA Grade should be stored at -20°C.