IGEPAL∣r CA-630
IGEPAL∣r CA-630
IGEPAL∣r CA-630
IGEPAL∣r CA-630
Thermo Scientific Chemicals

IGEPAL∣r CA-630

IGEPAL∣r CA-630, CAS # 9002-93-1, is a nonionic and non-denaturing detergent used mainly for the solubilization, isolation, and purification of membrane protein complexes.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61055.AP
also known as J61055-AP
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS9002-93-1
IUPAC Name2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
InChI KeyJYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILESCC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Synonymtriton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy
FormViscous liquid
Chromatographic Purity63.0 to 67.0° (1% inwater)
Cloud point63.0 to 67.0° (1% inwater)
Water Content (Karl Fischer Titration)≤0.50%
Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins, IGEPAL™ CA-630 is used as a detergent in paper industry, hand cleaners, metal cleaners. It is suitable for solubilization, isolation and purification of membrane protein complexes and in two-dimensional gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • IGEPAL|r CA-630 is a nonionic and non-denaturing detergent similar to Nonidet P-40

Application

  • IGEPAL|r CA-630 is mainly used for the solubilization, isolation, and purification of membrane protein complexes and in two-dimensional gel electrophoresis
  • IGEPAL|r CA-630 can be used in the composition of homogenizing buffers during western blotting technique in mouse muscle homogenates
  • It has been used in X-gal staining to study the lacZ expression
  • IGEPAL|r CA-630 is also a component of the cell lysis buffer in order to prepare RT-qPCR-ready cell lysates
RUO – Research Use Only

General References:

  1. Bertin, F. R.; Lemarié, C. A.; Robins, R. S.; Blostein, M. D. Growth arrest-specific 6 regulates thrombin-induced expression of vascular cell adhesion molecule-1 through forkhead box O1 in endothelial cells. J. Thromb. Haemost. 2015, 13 (12), 2260-2272.
  2. Blackford, A. N.; Nieminuszczy, J.; Schwab, R. A.; Galanty, Y.; Jackson, S. P.; Niedzwiedz, W. TopBP1 Interacts with BLM to Maintain Genome Stability but Is Dispensable for Preventing BLM Degradation. Mol. Cell 2015, 57 (6), 1133-1141.