Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, 99.9%
Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, 99.9%
Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, 99.9%
Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, 99.9%
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, 99.9%

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number J61062.36
also known as J61062-36
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Protease activityNone detected
RNaseNone detected
Assay from Supplier's CofA≥99.9%
DNaseNone detected
Insoluble material≤0.005%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only