Castanospermine, 99%
Castanospermine, 99%
Castanospermine, 99%
Thermo Scientific Chemicals

Castanospermine, 99%

Potent alpha and beta-glucosidase inhibitor | CAS: 79831-76-8 | C8H15NO4 | 189.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number J61071.ME
also known as J61071-ME
Price (USD)
720.00
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
720.00
Each
Specifications
Beilstein3588654
CAS79831-76-8
ChEBICHEBI:27860
Chemical Name or MaterialCastanospermine
ColorWhite
View more
Castanospermine, is used as a glycosidase inhibitor and antiinflammatory agent.castanospermine is used to inhibit syncytium formation between HIV-infected and CD4-expressing cells and may also interfere with infectivity. It has also been demonstrated that this agent inhibits inflammation at the level of leukocyte extravasation in rat models of experimental adjuvant-induced arthritis and autoimmune encephalomyelitis. Castanospermine is an inhibitor of Glucosidase I, Maltase-glucoamylase, and MANBA. Potent inhibitor of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids). Inhibits HIV syncytium formation and replication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Castanospermine, is used as a glycosidase inhibitor and antiinflammatory agent.castanospermine is used to inhibit syncytium formation between HIV-infected and CD4-expressing cells and may also interfere with infectivity. It has also been demonstrated that this agent inhibits inflammation at the level of leukocyte extravasation in rat models of experimental adjuvant-induced arthritis and autoimmune encephalomyelitis. Castanospermine is an inhibitor of Glucosidase I, Maltase-glucoamylase, and MANBA. Potent inhibitor of α- and β-glucosidases, especially glucosidase l (required for glucoprotein processing by transfer of mannose and glucose from asparagine-linked lipids). Inhibits HIV syncytium formation and replication.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Saul et al. Studies on the mechanism of castanospermine inhibition of α- and β-glucosidases. Arch.Biochem.Biophys. 1984, 230,(2), 668-75.
  2. Gruters et al. Interference with HIV-induced syncytium formation and viral infectivity by inhibitors of trimming glucosidase. Nature. 1987, 330,(6143), 74-7.