Insulin B (22-25) - FAQs
Insulin B (22-25) - FAQs
View additional product information for Insulin B (22-25) - FAQs (J61170.MB)
1 product FAQs found
What are insulin analogs?
There are 4 kinds of insulin analogs:
  • NPH insulin (Humulin N)
  • Lispro insulin (Humalog)
  • Insulin Aspart (Novolog)
  • Insulin glulisine