Brucine
Brucine
Brucine
Thermo Scientific Chemicals

Brucine

CAS: 357-57-3 | C23H26N2O4 | 394.471 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number J61178.06
also known as J61178-06
Price (USD)
30.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
30.10
Each
Chemical Identifiers
CAS357-57-3
IUPAC Name(1R,18S,20R,21R)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one
Molecular FormulaC23H26N2O4
InChI KeyRRKTZKIUPZVBMF-PLNGPGDESA-N
SMILESCOC1=CC2=C(C=C1OC)[C@@]13CCN4CC5=CCOC6CC(=O)N2C1[C@H]6[C@H]5C[C@@H]34
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated Ash1.0% max.
Assay (unspecified)98.0% min.
Brucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brucine is used as a denaturing alcohol, lubricant additive, separation of racemic mixtures. Proposed to reduce inflammatory mediators including COX-2, PGE2 and thromboxane B2. Able to induce cell apoptosis, with cytotoxic and antiproliferative effects.

Solubility
Soluble in alcohol, chloroform, and benzene, slightly soluble in water, ether, and glycerol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
Agrawal SS et al. Cytotoxic and antitumor effects of brucine on Ehrlich ascites tumor and human cancer cell line Life Sci. 2011, 89,(5-6), 147-58.Deng X et al. The apoptotic effect of brucine from the seed of Strychnos nux-vomica on human hepatoma cells is mediated via Bcl-2 and Ca2+ involved mitochondrial pathway. Toxicol Sci. 2006, 91,(1), 59-69.