TAPS sodium salt, 98%
TAPS sodium salt, 98%
TAPS sodium salt, 98%
Thermo Scientific Chemicals

TAPS sodium salt, 98%

Zwitterionic buffer | CAS: 91000-53-2 | C7H16NNaO6S | 265.256 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J61189.22
also known as J61189-22
Price (USD)
92.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
92.10
Each
Chemical Identifiers
CAS91000-53-2
IUPAC Namesodium 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonate
Molecular FormulaC7H16NNaO6S
InChI KeyFEGYIWVHCSRXCG-UHFFFAOYSA-M
SMILES[Na+].OCC(CO)(CO)NCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH10.0-11.0 (1% in water)
Appearance (Color)White
Assay from Supplier's CofA≥97.5%
FormPowder
Water Content (Karl Fischer Titration)≤2.0%
TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

Solubility
Soluble in water (333.33 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A.L.Flourat; A.A.M. Peru; A.R.S. Teixeira; F.Brunissen; F.Allais. Chemo-enzymatic synthesis of key intermediates (S)-γ-hydroxymethyl-α,β-butenolide and (S)-γ-hydroxymethyl-γ-butyrolactone via lipase-mediated Baeyer-Villiger oxidation of levoglucosenone. Green Chem. 2015, 17,(1), 404-412.
  2. Luboslav Starka; Martin Hill; Richard Hampl; Marie-Irene Malewiak; Aida Benalycherif; Robert Morfin; Jaroslav Kolena; Sona Scsukova. Studies on the mechanism of antiglucocorticoid action of 7α-hydroxydehydroepiandrosterone. Collect. Czech. Chem. Commun. 1998, 63,(10), 1683-1698.