Product Image
Thermo Scientific Chemicals

Acrylamide/Bisacrylamide 37.5:1, powder

Premixed powder for preparing nucleic acid PAGE gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
40 g
200 g
Catalog number J61220.17
also known as J61220-17
Price (USD)
64.65
Online exclusive
71.50
Save 6.85
Each
Quantity:
40 g
Request bulk or custom format
Price (USD)
64.65
Online exclusive
71.50
Save 6.85
Each
Specifications
Chemical Name or MaterialAcrylamide/Bisacrylamide 37.5:1
DOT InformationTransport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ACRYLAMIDE, SOLID
Health Hazard 1H301-H312+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H301-H340-H350-H372-H361-H312-H332-H315-H319-H317
Toxic if swallowed.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
View more
Premixed powder for preparing nucleic acid PAGE gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only