What is U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) soluble in?
U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) is soluble in DMSO. It is poorly soluble in ethanol and water.
Search
View additional product information for U0126, 99+% - FAQs (J61246.MC, J61246.MB)
1 product FAQs found