Vasopressin, 98+%
Vasopressin, 98+%
Vasopressin, 98+%
Thermo Scientific Chemicals

Vasopressin, 98+%

CAS: 11000-17-2 | C46H65N15O12S2
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J61248.MC
also known as J61248-MC
Price (USD)
1,222.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,438.00
Save 215.35 (15%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
1,222.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,438.00
Save 215.35 (15%)
Each
Chemical Identifiers
CAS11000-17-2
SpecificationsSpecification SheetSpecification Sheet
Purity≥98.0% (LCMS)
Assay from Suppliers CofAPeptide Content: ≥70.0%
FormSolid
LCMSConforms
Appearance (Color)White to off-white
It is used as a peptide hormone that regulates the body’s retention of water. Vasopressin or antidiuretic hormone is a potent endogenous hormone which is responsible for regulating plasma osmolality and volume. It acts as a neurotransmitter in the brain to control circadian rhythm, thermoregulation, and adrenocorticotrophic hormone release (ACTH). The therapeutic use of vasopressin has become increasingly important in the critical care environment in the management of cranial diabetes insipidus, bleeding abnormalities, oesophageal variceal haemorrhage, asystolic cardiac arrest, and septic shock.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (Miscible).

Notes
Store in airtight containers at 2° to 8°. Store in cool, sealed container.
RUO – Research Use Only

General References:

  1. Andrew Sharman.; James Low. Vasopressin and its role in critical care. Contin Educ Anaesth Crit Care Pain . 2008, 8,(4), 134-137.
  2. LV Scott.; TG Dinan. Vasopressin and the regulation of hypothalamic-pituitary-adrenal axis function: implications for the pathophysiology of depression. Life sciences. 1998, 62,(22), 1985-1998.