Puromycin dihydrochloride, 98%
Puromycin dihydrochloride, 98%
Puromycin dihydrochloride, 98%
Puromycin dihydrochloride, 98%
Thermo Scientific Chemicals

Puromycin dihydrochloride, 98%

Puromycin dihydrochloride hydrate, CAS # 58-58-2, is an aminoglycoside showing antibiotic, anti-neoplastic, and anti-protozoal activities. This compound also has the ability to inhibit protein synthesis. | CAS: 58-58-2 | C22H31Cl2N7O5 | 544.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61278.MC
also known as J61278-MC
Price (USD)
274.65
Special offer
Online exclusive
Ends: 30-Jun-2026
323.00
Save 48.35 (15%)
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
274.65
Special offer
Online exclusive
Ends: 30-Jun-2026
323.00
Save 48.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS58-58-2
IUPAC Name(2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide dihydrochloride
Molecular FormulaC22H31Cl2N7O5
InChI KeyMKSVFGKWZLUTTO-MLYJQVMKNA-N
SMILESCl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Solution TestClear, colorless
Appearance (Color)White to pale cream
FormPowder
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)≤12.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Information

• Puromycin dihydrochloride hydrate is the hydrochloride salt form of puromycin and is an aminoglycoside with antibiotic activity isolated from the Streptomyces alboniger
• Puromycin dihydrochloride hydrate is a protein synthesis inhibitor that causes premature chain termination by acting as an analog of the 3’-terminal ends of aminoacyl-tRNA, inhibiting protein synthesis

Applications

• This compound shows anti-microbial, anti-protozoal, anti-trypanosomal, and anti-neoplastic activities and is also used as an antibiotic in cell culture
• It also reveals anti-tumor activity in studies on brain cancer cells
• This compound has been used to study transcriptional regulatory mechanisms that control the sequential and coordinate expression of genes during cell differentiation
• It is active against gram-positive microorganisms, less active against acid-fast bacilli and gram-negative microorganisms. Puromycin can prevent growth of bacteria, protozoa, algae, and mammalian cells in in vitro studies.

RUO – Research Use Only

General References:

  1. Rosson, R. A.; Tebo, B. M.; Nealson, K. H. Use of poisons in determination of microbial manganese binding rates in seawater. Appl. Environ. Microbiol. 1984, 47, (7), 740-745.
  2. Nair, V.; Emanuel, D. J.. Synthetic design, stereochemistry, and enzymic activity of a reversed aminoacyl nucleoside: an analog of puromycin. J. Am. Chem. Soc. 1977, 99, (5), 1571-1576.