(R)-(-)-Deprenyl hydrochloride
(R)-(-)-Deprenyl hydrochloride
(R)-(-)-Deprenyl hydrochloride
Thermo Scientific Chemicals

(R)-(-)-Deprenyl hydrochloride

Selective MAO-B inhibitor; anti-Parkinsonian agent | CAS: 14611-52-0 | C13H18ClN | 223.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61286.03
also known as J61286-03
Price (USD)
222.65
Special offer
Online exclusive
Ends: 30-Jun-2026
262.00
Save 39.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
222.65
Special offer
Online exclusive
Ends: 30-Jun-2026
262.00
Save 39.35 (15%)
Each
Chemical Identifiers
CAS14611-52-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Elemental AnalysisHydrogen: 7.46-8.76% (Theoretical 8.11%)
Assay from Supplier's CofA≥95% (HPLC)
FormPowder or crystalline powder or solid
Elemental AnalysisCarbon: 65.60-73.97% (Theoretical 69.79%)
View more
(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

Solubility
Soluble in water at 100mM

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. J.W.Tetrud; J.W.Langston. The effect of deprenyl (selegiline) on the natural history of Parkinson's disease. Science. 1989, 245,(4917), 519-522.
  2. E.W.Tsang; C.Hamani; E.Moro; F.Mazzella; U.Saha; A.M.Lozano; M.Hodaie; R.Chuang; T.Steeves; S.Y.Lim; B.Neagu; R.Chen. Subthalamic deep brain stimulation at individualized frequencies for Parkinson disease. Neurology. 2012, 78,(24), 1930-1938.