Amoxicillin trihydrate
Amoxicillin trihydrate
Amoxicillin trihydrate
Thermo Scientific Chemicals

Amoxicillin trihydrate

A semi-synthetic antibiotic similar to penicillin | CAS: 61336-70-7 | C16H25N3O8S | 419.449 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61290.06
also known as J61290-06
Price (USD)
46.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.00
Each
Chemical Identifiers
CAS61336-70-7
IUPAC Name(2S,5R,6R)-6-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
Molecular FormulaC16H25N3O8S
InChI KeyMQXQVCLAUDMCEF-CWLIKTDRSA-N
SMILESO.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=C(O)C=C3)C(=O)N2[C@H]1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content≤5.0 %
Water Content (Karl Fischer Titration)11.5-14.5%
Appearance (Color)White to pale cream
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
View more
Amoxicillin trihydratesemi-synthetic antibiotic related to Penicillin which has antibacterial poperties. It is used for treatment of a variety of bacterial infections. It is used as a pharmaceutical product as a antibiotic agent, anti-infective drug raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amoxicillin trihydratesemi-synthetic antibiotic related to Penicillin which has antibacterial poperties. It is used for treatment of a variety of bacterial infections. It is used as a pharmaceutical product as a antibiotic agent, anti-infective drug raw materials.

Solubility
Soluble in water at 3.4mg/ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
RUO – Research Use Only

General References:

  1. SK Yellanki.; J Singh.; JA Syed.; R Bigala. Design and characterization of amoxicillin trihydrate mucoadhesive microspheres for prolonged gastric retention. International Journal of Pharmaceutical Sciences and Drug Research. 2010, 2, (2), 112-114.
  2. AK Hilton.; PB Deasy. Use of hydroxypropyl methylcellulose acetate succinate in an enteric polymer matrix to design controlled-release tablets of amoxicillin trihydrate. Journal of Pharmaceutical Sciences. 1993, 82, (7), 737-743.