Carboxy-PTIO potassium salt, 98+%
Carboxy-PTIO potassium salt, 98+%
Carboxy-PTIO potassium salt, 98+%
Thermo Scientific Chemicals

Carboxy-PTIO potassium salt, 98+%

Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase | CAS: 148819-94-7 | C14H17KN2O4 | 316.398 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Catalog number J61316.MA
also known as J61316-MA
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
133.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS148819-94-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.0% min.
Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only