Busulfan, 98%
Busulfan, 98%
Busulfan, 98%
Thermo Scientific Chemicals

Busulfan, 98%

A cell cycle non-specific alkylating antineoplastic agent | CAS: 55-98-1 | C6H14O6S2 | 246.292 g/mol
Have Questions?
Catalog NumberQuantity
J61348.06
also known as J61348-06
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61348.06
also known as J61348-06
Price (USD)
31.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
31.10
Each
Specifications
Beilstein1791786
CAS55-98-1
ChEBICHEBI:28901
Chemical Name or MaterialBusulfan
ColorBrown
View more
A cell cycle non-specific alkylating antineoplastic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only