This product is animal origin and can only be shipped within the United States
Clostripain, Clostridium histolyticum, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Clostripain, Clostridium histolyticum, Thermo Scientific Chemicals

A protease that cleaves proteins on the carboxyl bond of arginine
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
10 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61362.MCR
also known as J61362-M^
Price (USD)
-
Quantity:
1 mg
Request bulk or custom format
Specifications
CAS9028-00-6
Chemical Name or MaterialClostripain
ColorWhite
EINECS Number232-822-2
Health Hazard 1H315-H319-H334-H335
View more
A protease that cleaves proteins on the carboxyl bond of arginine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

How many units of clostripain are in one milligram (dry weight) of Clostripain, Clostridium histolyticum?

Each mg of Clostripain, Clostridium histolyticum (dry weight) contains a minimum of 50.0 units of clostripain.

Does Clostripain, Clostridium histolyticum, contain a His Tag?

The clostripain in Clostripain, Clostridium histolyticum is the native protein and does not have a His Tag.