Bezafibrate, 98+%
Bezafibrate, 98+%
Bezafibrate, 98+%
Thermo Scientific Chemicals

Bezafibrate, 98+%

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent | CAS: 41859-67-0 | C19H20ClNO4 | 361.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61412.03
also known as J61412-03
Price (USD)
39.65
Online exclusive
44.30
Save 4.65
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Online exclusive
44.30
Save 4.65
Each
Specifications
CAS41859-67-0
ChEBICHEBI:47612
Chemical Name or MaterialBezafibrate
ColorWhite
EINECS Number255-567-9
View more
A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only