Furosemide, 97+%
Furosemide, 97+%
Furosemide, 97+%
Thermo Scientific Chemicals

Furosemide, 97+%

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter | CAS: 54-31-9 | C12H11ClN2O5S | 330.739 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61457.14
also known as J61457-14
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Specifications
CAS54-31-9
ChEBICHEBI:47426
Chemical Name or MaterialFurosemide
ColorWhite
EINECS Number200-203-6
View more
Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only