Luminol and oxidizing solutions (each bottle)
Thermo Scientific Chemicals

Luminol and oxidizing solutions (each bottle)

For chemiluminecent detection of HRP-based Western blotting systems
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61462.AK
also known as J61462-AK
Price (USD)
188.00
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
188.00
Each
Specifications
Chemical Name or MaterialLuminol and oxidizing solutions (each bottle)
Physical FormLiquid
Recommended StorageKeep cold
SensitivityLight sensitive
Solubility InformationIt is soluble in most polar organic solvents but is insoluble in water.
View more
Used by biologists in cellular assays for the detection of copper, iron, and cyanides, as well as of specific proteins by western blot. For chemiluminecent HRP-based detection of western blotting systems. Microestimation of glucose and glucose oxidase using enzyme-induced chemiluminescence6,7 - Inhibitor of poly ADP Ribose Synthase8 - Demonstrations of the phenomenon of chemiluminescence, using different sensitizers to produce different colors of light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only