4-Aminopyridine, 99+%
4-Aminopyridine, 99+%
4-Aminopyridine, 99+%
Thermo Scientific Chemicals

4-Aminopyridine, 99+%

A potassium channel blocker | CAS: 504-24-5 | C5H6N2 | 94.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61470.22
also known as J61470-22
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS504-24-5
IUPAC Namepyridin-4-amine
Molecular FormulaC5H6N2
InChI KeyNUKYPUAOHBNCPY-UHFFFAOYSA-N
SMILESNC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.0%
FormPowder or crystals or crystalline powder
A potassium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only