BioAcryl-P (30%, 37.5:1) - FAQs
BioAcryl-P (30%, 37.5:1) - FAQs
View additional product information for BioAcryl-P (30%, 37.5:1) - FAQs (J61505.K2, J61505.AP)
2 product FAQs found
What are the recommend storage conditions for BioAcryl-P (30%, 37.5:1) (Cat. No. J61505.K2, J61505-K2)?
We recommend storing BioAcryl-P (30%, 37.5:1) at 2-8 degrees C.
What is the composition of BioAcryl-P (30%, 37.5:1)?
BioAcryl-P (30%, 37.5:1) contains, 300 g acrylamide, 8 g bis-acryamide, in ddH2O to 1 L. The 30% represents the weight by volume of both acrylamide and bis-acrylamide (308 g/1,000 mL or~30.8%). The ratio 37.5:1 is the w/w ratio of acrylamide:bis-acrylamide in the solution (300 g:8 g or 37.5:1).