Methyl Green, zinc chloride
Methyl Green, zinc chloride
Methyl Green, zinc chloride
Thermo Scientific Chemicals

Methyl Green, zinc chloride

CAS: 7114-03-6 | C27H35Cl4N3Zn | 608.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J61509.22
also known as J61509-22
Price (USD)
362.65
Online Exclusive
403.00
Save 40.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
362.65
Online Exclusive
403.00
Save 40.35 (10%)
Each
Specifications
Beilstein3914106
CAS7114-03-6
Chemical Name or MaterialMethyl Green, zinc chloride
EINECS Number230-415-4
Formula Weight653.24
View more
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

Solubility
Soluble in water, ethanol or methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. N.B. Kurnick. The quantitative estimation of desoxyribosenucleic acid based on methyl green staining. Experimental Cell Research. 1950, 1 (2), 151-158.
  2. Seog K. Kim.; Bengt Norden. Methyl green: A DNA major-groove binding drug. Febs letters. 1993, 315, (1), 61-64.