Bradford Dye Reagent, Ready-to-Use soln. - FAQs
Bradford Dye Reagent, Ready-to-Use soln. - FAQs
View additional product information for Bradford Dye Reagent, Ready-to-Use soln. - FAQs (J61522.K2, J61522.AP)
1 product FAQs found
At what temperature should I store Bradford Dye Reagent, Ready-to-Use soln.?
The Bradford Dye Reagent, Ready-to-Use soln. should be stored refrigerated at 2-8 degrees C.