(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals
(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals
(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific™

(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J61591.LB0
also known as J61591-#0
Price (USD)
229.65
Special offer
Online exclusive
Ends: 30-Jun-2026
270.00
Save 40.35 (15%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
229.65
Special offer
Online exclusive
Ends: 30-Jun-2026
270.00
Save 40.35 (15%)
Each
Specifications
Beilstein1078734
CAS52809-07-1
ChEBICHEBI:8734
Chemical Name or Material(+)-Quisqualic acid
ColorWhite
View more
Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.

Solubility
Soluble in ethanol, water. Insoluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only