Emodin
Emodin
Emodin
Thermo Scientific Chemicals

Emodin

A protein tyrosine kinase inhibitor | CAS: 518-82-1 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61600.03
also known as J61600-03
Price (USD)
696.65
Online Exclusive
774.00
Save 77.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
696.65
Online Exclusive
774.00
Save 77.35 (10%)
Each
Chemical Identifiers
CAS518-82-1
IUPAC Name1,3,8-trihydroxy-6-methyl-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC15H10O5
InChI KeyRHMXXJGYXNZAPX-UHFFFAOYSA-N
SMILESCC1=CC(O)=C2C(=O)C3=C(O)C=C(O)C=C3C(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying1.0% max.
Appearance (Color)Pale brown
FormPowder
Assay (HPLC)90.0% min.
Ash1.0% max.
A protein tyrosine kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only