Aniracetam
Aniracetam
Aniracetam
Thermo Scientific Chemicals

Aniracetam

CAS: 72432-10-1 | C12H13NO3 | 219.24 g/mol
Have Questions?
Catalog NumberQuantity
J61661.03
also known as J61661-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61661.03
also known as J61661-03
Price (USD)
111.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
111.00
Each
Specifications
CAS72432-10-1
ChEBICHEBI:47943
Chemical Name or MaterialAniracetam
ColorWhite to Yellow
Formula Weight219.24
View more
AMPA receptor potentiator. Slows the rate of ion-channel closing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • AMPA receptor potentiator
  • Slows the rate of ion-channel closing
RUO – Research Use Only