Lecithin, 90%, soybean
Lecithin, 90%, soybean
Lecithin, 90%, soybean
Thermo Scientific Chemicals

Lecithin, 90%, soybean

CAS: 8002-43-5 | C42H80NO8P | 758.075 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
250 g
Catalog number J61675.18
Price (USD)
25.60
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
25.60
Each
Chemical Identifiers
CAS8002-43-5
IUPAC Name(2-{[3-(hexadecanoyloxy)-2-(octadeca-9,12-dienoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular FormulaC42H80NO8P
InChI KeyJLPULHDHAOZNQI-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)max. 1.0%
Peroxide contentmax. 5.0 meq/kg
Assay (unspecified)Acetone insolubles: min. 97%
Assay (unspecified)Hexane insolubles: max. 0.3%
Acid Valuemax. 35 mg KOH/g
View more
Lecithin is used as a dietary supplement, food ingredient, and food additive. It is also used as a liquid carrier for plastics additives, dyes or pigments. Its primary functions are as an emulsifier, antioxidant, and release agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lecithin is used as a dietary supplement, food ingredient, and food additive. It is also used as a liquid carrier for plastics additives, dyes or pigments. Its primary functions are as an emulsifier, antioxidant, and release agent.

Solubility
Partly soluble in water, partly soluble in acetone, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. A Smiles.; Y Kakuda.; BE MacDonald. Effect of degumming reagents on the recovery and nature of lecithins from crude canola,soybean and sunflower oils. Journal of the American Oil Chemists’ Society. 1988, 65, (7), 1151-1155.
  2. L Rydhag.; I Wilton. The function of phospholipids of soybean lecithin in emulsions. Journal of the American Oil Chemists’ Society. 1981, 58, (8), 830-837.