Phenolphthalein, 2% soln. in 95% ethanol
Phenolphthalein, 2% soln. in 95% ethanol
Phenolphthalein, 2% soln. in 95% ethanol
Thermo Scientific Chemicals

Phenolphthalein, 2% soln. in 95% ethanol

pH indicator | CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1000 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61785.AE
also known as J61785-AE
Price (USD)
28.30
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
28.30
Each
Chemical Identifiers
CAS77-09-8
IUPAC Name3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC20H14O4
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (special)Visual transition interval: Passes test
FormLiquid
It is used as a laboratory reagent and pH indicator. Phenolphthalein is a common indicator to use in neutralization titrations. Phenolphthalein exerts laxative effects by stimulating the intestinal mucosa and constricting smooth muscles. It is used medicinally as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only