MOPS, 1.0M buffer soln., pH 7.0
MOPS, 1.0M buffer soln., pH 7.0
MOPS, 1.0M buffer soln., pH 7.0
Thermo Scientific Chemicals

MOPS, 1.0M buffer soln., pH 7.0

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61821.AK
also known as J61821-AK
Price (USD)
151.65
Online Exclusive
168.00
Save 16.35 (10%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
151.65
Online Exclusive
168.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS1132-61-2
IUPAC Name4-(3-sulfonatopropyl)morpholin-4-ium
Molecular FormulaC7H15NO4S
InChI KeyDVLFYONBTKHTER-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)CCC[NH+]1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
pH7.0 ± 0.15
CommentDissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through 0.22 micron filter
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
RUO – Research Use Only
MM Santoro.; DW Bolen. A test of the linear extrapolation of unfolding free energy changes over an extended denaturant concentration range. Biochemistry. 1992, 31, (20), 4901-4907.