MOPS, 1.0M buffer soln., pH 7.5
MOPS, 1.0M buffer soln., pH 7.5
MOPS, 1.0M buffer soln., pH 7.5
MOPS, 1.0M buffer soln., pH 7.5
Thermo Scientific Chemicals

MOPS, 1.0M buffer soln., pH 7.5

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61843.AP
also known as J61843-AP
Price (USD)
210.65
Special offer
Online exclusive
Ends: 30-Jun-2026
248.00
Save 37.35 (15%)
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
210.65
Special offer
Online exclusive
Ends: 30-Jun-2026
248.00
Save 37.35 (15%)
Each
Add to cart
Specifications
CAS1132-61-2
ChEBICHEBI:44115
Chemical Name or MaterialMOPS buffer soln.
ColorColorless
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only