Protease Inhibitor Cocktail, for general use
Thermo Scientific Chemicals
Protease Inhibitor Cocktail, for general use
Have Questions?
Catalog NumberQuantity
J61852.XF
also known as J61852-XF
1 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61852.XF
also known as J61852-XF
Price (USD)
-
Quantity:
1 mL
Request bulk or custom format
Specifications
OdorOdorless
Quantity1 mL
Recommended StorageKeep cold
FormLiquid
Product TypeProtease Inhibitor Cocktail
View more
Phosphatase Inhibitor Cocktail is used to inhibitors with a broad specificity for serine, cysteine, and acid proteases, and aminopeptidases. It has been tested on a wide variety of cells and tissue extracts to isolate/maintain intact proteins without degradation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C