D-Lactic acid
D-Lactic acid
D-Lactic acid
Thermo Scientific Chemicals

D-Lactic acid

Useful chiral synthon, building block for depsipeptides | CAS: 10326-41-7 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J61871.03
Price (USD)
797.65
Online exclusive
886.00
Save 88.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
797.65
Online exclusive
886.00
Save 88.35
Each
Chemical Identifiers
CAS10326-41-7
IUPAC Name2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILESCC(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Impurity contentL Enantiomer: ≤0.2% (Chiral chromatography, vendor CofA)
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥99.0% (TLC)
Water Content (Karl Fischer Titration)≤1.0%
D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.

Solubility
Soluble in water (50mg/ml)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

General References:

  1. Kobayashi D.; Nozawa T.; Imai K.; Nezu J.; Tsuji A.; Tamai I. Involvement of human organic anion transporting polypeptide OATP-B (SLC21A9) in pH-dependent transport across intestinal apical membrane. J Pharmacol Exp Ther. 2003, 306, (2), 703-8.
  2. Nozawa T.; Imai K.; Nezu J.; Tsuji A.; Tamai I. Functional characterization of pH-sensitive organic anion transporting polypeptide OATP-B in human. J Pharmacol Exp Ther. 2003, 308, (2), 438-45.