What is the molecular weight of Alginic Acid sodium salt, high viscosity (Cat. No. J61887.xx)?
Based on the viscosity of the chemical (1335 mPa= 1335 cps), the approximate molecular weight of Alginic Acid sodium salt, high viscosity is 120,000 Da.
Search
View additional product information for Alginic acid sodium salt, high viscosity - FAQs (J61887.22, J61887.A1, J61887.30)
1 product FAQs found