Lithium chloride, 1M aq. soln.
Lithium chloride, 1M aq. soln.
Lithium chloride, 1M aq. soln.
Thermo Scientific Chemicals

Lithium chloride, 1M aq. soln.

CAS: 7447-41-8 | ClLi | 42.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
Catalog number J61895.AE
also known as J61895-AE
Price (USD)
88.90
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
88.90
Each
Specifications
CAS7447-41-8
ChEBICHEBI:48607
Chemical Name or MaterialLithium chloride
ColorColorless
Concentration or Composition (by Analyte or Components)1M aq. soln.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only