Quinapril hydrochloride, 98%
Quinapril hydrochloride, 98%
Quinapril hydrochloride, 98%
Thermo Scientific Chemicals

Quinapril hydrochloride, 98%

Angiotensin-converting enzyme inhibitor | CAS: 82586-55-8 | C25H31ClN2O5 | 474.982 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61913.MC
also known as J61913-MC
Price (USD)
52.50
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
52.50
Each
Specifications
CAS82586-55-8
ChEBICHEBI:8714
Chemical Name or MaterialQuinapril hydrochloride
ColorWhite
Formula Weight474.99
View more
Angiotensin-converting enzyme inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only