What are the concentrations of the components in Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula?
Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula contains 89 mM sodium phosphate dibasic, 15 mM potassium phosphate monobasic, 1.37 mM sodium chloride, and 27 mM potassium chloride.