Amido Black 10B, 0.2% v/v soln. in 5% acetic acid
Amido Black 10B, 0.2% v/v soln. in 5% acetic acid
Amido Black 10B, 0.2% v/v soln. in 5% acetic acid
Thermo Scientific Chemicals

Amido Black 10B, 0.2% v/v soln. in 5% acetic acid

CAS: 1064-48-8 | C22H14N6Na2O9S2 | 616.487 g/mol
Have Questions?
Catalog NumberQuantity
J61978.AP
also known as J61978-AP
500 mL
Catalog number J61978.AP
also known as J61978-AP
Price (USD)
90.50
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
90.50
Each
Specifications
Beilstein5374263
CAS1064-48-8
Chemical Name or MaterialAmido Black 10B
Concentration0.2% v/v soln. in 5% acetic acid
Formula Weight616.49
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only