Tamsulosin hydrochloride, 98+%
Tamsulosin hydrochloride, 98+%
Tamsulosin hydrochloride, 98+%
Thermo Scientific Chemicals

Tamsulosin hydrochloride, 98+%

An alpha-1 adrenoceptor antagonist | CAS: 106463-17-6 | C20H29ClN2O5S | 444.971 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
25 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61999.MA
also known as J61999-MA
Price (USD)
120.65
Online exclusive
134.00
Save 13.35
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
120.65
Online exclusive
134.00
Save 13.35
Each
Chemical Identifiers
CAS106463-17-6
IUPAC Namehydrogen 5-[(2R)-2-{[2-(2-ethoxyphenoxy)ethyl]amino}propyl]-2-methoxybenzene-1-sulfonamide chloride
Molecular FormulaC20H29ClN2O5S
InChI KeyZZIZZTHXZRDOFM-XFULWGLBSA-N
SMILES[H+].[Cl-].CCOC1=CC=CC=C1OCCN[C@H](C)CC1=CC=C(OC)C(=C1)S(N)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream
FormPowder
Assay from Suppliers CofA≥98.0% (UHPLC)
Loss on Drying≤0.5%
View more
An alpha-1 adrenoceptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only