Phosphate-buffered saline (PBS, 10X), pH 7.4 - FAQs
Phosphate-buffered saline (PBS, 10X), pH 7.4 - FAQs
View additional product information for Phosphate-buffered saline (PBS, 10X), pH 7.4 - FAQs (J62036.K7, J62036.K3, J62036.K2)
1 product FAQs found
What is the composition of Phosphate-buffered saline (PBS, 10X), pH 7.4 (Cat. No. J62036.K7, J62036-K7, J62036.K2, J62036-K2, J62036.K3, J62036-K3)?
Here is the composition of Phosphate-buffered saline (PBS, 10X), pH 7.4 (Cat. No. J62036.K7, J62036-K7, J62036.K2, J62036-K2, J62036.K3, J62036-K3):
- Sodium chloride: 1.37 M
- Potassium chloride (KCl): 27 mM
- Phosphate buffer: 101.4 mM
- Potassium phosphate monobasic: 17.6 mM