Diosmin
Diosmin
Diosmin
Thermo Scientific Chemicals

Diosmin

CAS: 520-27-4 | C28H32O15 | 608.549 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62073.06
also known as J62073-06
Price (USD)
40.50
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.50
Each
Specifications
CAS520-27-4
ChEBICHEBI:4631
Chemical Name or MaterialDiosmin
ColorBrown to Yellow
EINECS Number208-289-7
View more
Diosmin exhibits anti-proliferative and anti-inflammatory. It is used in the treatment of venous disease, i.e., chronic venous insufficiency (CVI) and hemorrhoidal disease (HD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO (50 mg/ml), water (122 mg/ml at 25°C), and ethanol (<1mg/mL at 25°C).

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
RUO – Research Use Only

General References:

  1. Chung-Hsi Chou; Cheng-Wei Liu; Deng-Jye Yang; Yi-Hsieng Samuel Wu; Yi-Chen Chen. Amino acid, mineral, and polyphenolic profiles of black vinegar, and its lipid lowering and antioxidant effects in vivo. Food Chemistry.2003, 168,
  2. Oomaidurai Senthamizhselvan; Jeganathan Manivannan; Thangarasu Silambarasan; Boobalan Raja. Diosmin pretreatment improves cardiac function and suppresses oxidative stress in rat heart after ischemia/reperfusion. European Journal of Pharmacology.2009, 736,