TRIS, 1.0M buffer soln., pH 9.5
TRIS, 1.0M buffer soln., pH 9.5
TRIS, 1.0M buffer soln., pH 9.5
Thermo Scientific Chemicals

TRIS, 1.0M buffer soln., pH 9.5

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantity
J62084.K2
also known as J62084-K2
1 L
Catalog number J62084.K2
also known as J62084-K2
Price (USD)
91.65
Special offer
Online exclusive
Ends: 30-Jun-2026
108.00
Save 16.35 (15%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
91.65
Special offer
Online exclusive
Ends: 30-Jun-2026
108.00
Save 16.35 (15%)
Each
Specifications
CAS77-86-1
ChEBICHEBI:9754
Chemical Name or MaterialTRIS buffer soln.
ColorColorless
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
View more
TRIS, 1.0M buffer soln., pH 9.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TRIS, 1.0M buffer soln., pH 9.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

Solubility
Soluble in water and organic solvents.

Notes
Store in cool place. Keep container
RUO – Research Use Only

General References:

  1. DE Soltis.; CH Haufler.; DC Darrow.; GJ Gastony. Starch gel electrophoresis of ferns: a compilation of grinding buffers, gel and electrodebuffers, and staining schedules. American fern journal. 1983, 73 ,(1),9-27.
  2. DE Soltis.; CH Haufler.; DC Darrow.; GJ Gastony. Starch gel electrophoresis of ferns: a compilation of grinding buffers, gel and electrodebuffers, and staining schedules. J. Appl. Cryst.1991, 24 ,409-411.