Hesperidin, 95%
Hesperidin, 95%
Hesperidin, 95%
Thermo Scientific Chemicals

Hesperidin, 95%

Acts as an antioxidant | CAS: 520-26-3 | C28H34O15 | 610.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62126.14
also known as J62126-14
Price (USD)
61.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
61.60
Each
Chemical Identifiers
CAS520-26-3
IUPAC Name(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC28H34O15
InChI KeyQUQPHWDTPGMPEX-QJBIFVCTSA-N
SMILESCOC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0% (HPLC)
Appearance (Color)White to pale yellow
Water Content (Karl Fischer Titration)≤5.0%
FormPowder
Heavy Metals≤20 ppm
Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hesperidin which has been found to be a potent chemopreventive agent in oral, urinary bladder, and colon carcinogenesis in animal models. Applied in food field, it is used as a flavour enhancer and sweetener in a wide variety of alcoholicbeverage, dessert foods and savory foods, applied in cosmetics, it can be added into toothpaste and mouth wash, applied in pharmaceutical field, it is mainly used to reducing the bitterness of in form. It also enhance the action of Vitamin C.

Solubility
Insoluble in water. Soluble in organic solvents such as DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents. Keep away from strong light and heat.
RUO – Research Use Only

General References:

  1. Al-Ashaal HA & El-Sheltawy ST. Antioxidant capacity of hesperidin from citrus peel using electron spin resonance and cytotoxic activity against human carcinoma cell lines. Pharm Biol . 2011, 49, (3), 276-82.
  2. Hirata A et al. Kinetics of radical-scavenging activity of hesperetin and hesperidin and their inhibitory activity on COX-2 expression. Anticancer Res . 2011, 25, (5), 3367-74.