bis-Benzimide H-33342 trihydrochloride trihydrate, 98%
bis-Benzimide H-33342 trihydrochloride trihydrate, 98%
bis-Benzimide H-33342 trihydrochloride trihydrate, 98%
Thermo Scientific Chemicals

bis-Benzimide H-33342 trihydrochloride trihydrate, 98%

CAS: 23491-52-3 | C27H28N6O | 452.562 g/mol
Have Questions?
Catalog NumberQuantity
J62134.MC
also known as J62134-MC
100 mg
Catalog number J62134.MC
also known as J62134-MC
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Specifications
Beilstein4088183
CAS23491-52-3
ChEBICHEBI:51232
Chemical Name or Materialbis-Benzimide H-33342 trihydrochloride trihydrate
ColorYellow to Green
View more
Stain for identifying specific chromosome banding patterns

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only