Pancreatin, porcine pancreas
Thermo Scientific Chemicals

Pancreatin, porcine pancreas

Hydrolyzes starch into dextrins and sugar Pancreatin from porcine pancreas is used in the treatment of steatorrhea by lipase supplementation therapy and for pancreatic diabetes. It is also used in the removal of formalin-fixed tissues from arterial grafts without causing damage in fiber integrity.
Have Questions?
Change viewbuttonViewtableView
Quantity:
250g
1kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62162.30
also known as J62162-30
Price (USD)
-
Quantity:
250g
Request bulk or custom format
Chemical Identifiers
CAS8049-47-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Lipase activity: 2.0-10.0 x USP (Report USP u/mg)
CommentSource: Porcine pancreas
Assay (unspecified)Amylase activity: 1.0-2.5 x USP (Report USP u/mg)
Assay (unspecified)Protease activity: 1.0-2.5 x USP (Report USP u/mg)
FormPowder
Hydrolyzes starch into dextrins and sugar Pancreatin from porcine pancreas is used in the treatment of steatorrhea by lipase supplementation therapy and for pancreatic diabetes. It is also used in the removal of formalin-fixed tissues from arterial grafts without causing damage in fiber integrity. Further, it finds uses in contact lenses. In addition to this, it hydrolyzes starch into dextrins and sugar. It is also used to treat conditions responsible for deficiency in pancreatic secretions, such as surgical pancreatectomy, pancreatitis and cystic fibrosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only