Amlodipine besylate, 98+%
Amlodipine besylate, 98+%
Amlodipine besylate, 98+%
Thermo Scientific Chemicals

Amlodipine besylate, 98+%

An L-type calcium channel antagonist with potent antioxidant activity | CAS: 111470-99-6 | C26H31ClN2O8S | 567.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62164.03
also known as J62164-03
Price (USD)
109.65
Special offer
Online exclusive
Ends: 28-Dec-2026
129.00
Save 19.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
109.65
Special offer
Online exclusive
Ends: 28-Dec-2026
129.00
Save 19.35
Each
Specifications
CAS111470-99-6
ChEBICHEBI:2669
Chemical Name or MaterialAmlodipine besylate
ColorWhite
Formula Weight567.05
View more
Amlodipine besylate is used as a dihydropyridine calcium channel blocker. It is also used as an antianginal and antihypertensive. Amlodipine besylate also displays vasoprotective effects in cardiovascular disease and inhibits Ca2+-induced contractions in depolarized rat aorta (IC50 = 1.9 nM). Amlodipine besylate has been shown to inhibit proliferation of human vascular smooth muscle cells and epidermoid carcinoma A431 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amlodipine besylate is used as a dihydropyridine calcium channel blocker. It is also used as an antianginal and antihypertensive. Amlodipine besylate also displays vasoprotective effects in cardiovascular disease and inhibits Ca2+-induced contractions in depolarized rat aorta (IC50 = 1.9 nM). Amlodipine besylate has been shown to inhibit proliferation of human vascular smooth muscle cells and epidermoid carcinoma A431 cells.

Solubility
Soluble in DMSO (≥20 mg/mL), water (10 mM), chloroform, ethanol (≥14 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Burges et al. Calcium channel blocking properties of amlodipine in vascular smooth muscle and cardiac muscle in vitro: evidence for voltage modulation of vascular dihydropyridine receptors. J.Cardiovasc.Pharmacol. 1987, 9, ( 1), 110-9.
  2. Yoshida et al. Antiproliferative effect of Ca2+ channel blockers on human epidermoid carcinoma A431 cells. Eur.J.Pharmacol. 2003, 472, ( 1-2), 23-31.