L-Serine, Cell Culture Reagent
L-Serine, Cell Culture Reagent
L-Serine, Cell Culture Reagent
L-Serine, Cell Culture Reagent
Thermo Scientific Chemicals

L-Serine, Cell Culture Reagent

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
1 kg
Catalog number J62187.09
also known as J62187-09
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.70
Save 6.05 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.70
Save 6.05 (15%)
Each
Add to cart
Specifications
Beilstein1721404
CAS56-45-1
ChEBICHEBI:17115
Chemical Name or MaterialL-Serine
ColorWhite
View more
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

Solubility
Soluble in water.

Notes
Store in cool, dry place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Kenji Hashimoto.; Göran Engberg.; Eiji Shimizu.; Conny Nordin.; Leif H. Lindström.; Masaomi Iyo. Reduced d-serine to total serine ratio in the cerebrospinal fluid of drug naive schizophrenic patients.Progress in Neuro-Psychopharmacology and Biological Psychiatry. 2005, 29 (5),767-769 .
  2. Lekh Raj Juneja.; Toru Kazuoka.; Naomi Goto.;Tsuneo Yamane.; Shoichi Shimizu. Lekh Raj Juneja, Toru Kazuoka, Naomi Goto, Tsuneo Yamane *, Shoichi Shimizu.Biochimica et Biophysica Acta (BBA) - Lipids and Lipid Metabolism. 1989, 3 (28), 277-283.