(+/-)-Thalidomide, 99+%
(+/-)-Thalidomide, 99+%
(+/-)-Thalidomide, 99+%
Thermo Scientific Chemicals

(+/-)-Thalidomide, 99+%

CAS: 50-35-1 | C13H10N2O4 | 258.23 g/mol
Have Questions?
Catalog NumberQuantity
J62266.MC
also known as J62266-MC
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62266.MC
also known as J62266-MC
Price (USD)
119.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
119.00
Each
Chemical Identifiers
CAS50-35-1
IUPAC Name2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC13H10N2O4
InChI KeyUEJJHQNACJXSKW-UHFFFAOYNA-N
SMILESO=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Thin Layer Chromatography≥99.0% purity
Appearance (Color)White to cream
Infrared spectrumConforms
Assay from Supplier's CofA≥99.0% (HPLC)
FormPowder
Teratogenic compound found to inhibit HIV-1 replication and FGF-induced angiogenesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only