TE buffer, (20X, pH 7.6), RNAse free
TE buffer, (20X, pH 7.6), RNAse free
TE buffer, (20X, pH 7.6), RNAse free
Thermo Scientific Chemicals

TE buffer, (20X, pH 7.6), RNAse free

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
125 mL
250 mL
Catalog number J62285.EQE
also known as J62285-=E
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.30
Save 11.65 (15%)
Each
Quantity:
125 mL
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.30
Save 11.65 (15%)
Each
Specifications
CAS1185-53-1
Chemical Name or MaterialTE buffer
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
MDL NumberMFCD00236359
View more
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

Solubility
Fully miscible in water.

Notes
200mM Tris-HCl with 20mM EDTA
RUO – Research Use Only
Valerie S Latimer.; Sarina M Rodrigues.; Vasilios T Garyfallou.; Steven G Kohama.; Richard B White.; Russell D Fernald.; Henryk F Urbanski. Two molecular forms of gonadotropin-releasing hormone (GnRH-I and GnRH-II) are expressed by two separate populations of cells in the rhesus macaque hypothalamus.Molecular Brain Research. 2000, 75 (2),287-292 .