Phenol:Chloroform:Isoamyl alcohol  25:24:1, Ready-to-Use saturated aq. soln., pH 5.2
Phenol:Chloroform:Isoamyl alcohol  25:24:1, Ready-to-Use saturated aq. soln., pH 5.2
Phenol:Chloroform:Isoamyl alcohol  25:24:1, Ready-to-Use saturated aq. soln., pH 5.2
Thermo Scientific Chemicals

Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 5.2

Phenol-Chloroform-Isoamyl alcohol mixture in the ratio of 25:24:1 (v/v/v), CAS # 136112-00-0, 67-66-3, 123-51-3, is a solution used in the purification of nucleic acids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
J62336.AE
also known as J62336-AE
100mL
J62336.AN
also known as J62336-AN
400mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62336.AE
also known as J62336-AE
Price (USD)
-
Quantity:
100mL
Request bulk or custom format
Chemical Identifiers
CAS136112-00-0
IUPAC Name3-methylbutan-1-ol; phenol; trichloromethane
Molecular FormulaC12H19Cl3O2
InChI KeyZYWFEOZQIUMEGL-UHFFFAOYSA-N
SMILESClC(Cl)Cl.CC(C)CCO.OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance atOD at 330nm: ≤ 0.2; at 405nm: ≤ 0.1; at 510nm: ≤ 0.1
Assay (special)≥99.0%
pH4.9 to 5.5
Appearance (Color)Clear colorless
FormLiquid
Phenol:Chloroform:Isoamyl alcohol 25:24:1 is used in the extraction of DNA for the generation of DNA minisatellite fingerprints. It is also used for purifying RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• This solution contains a mixture of pure chloroform, isoamyl alcohol and phenol saturated with Tris-EDTA, with a stabilized pH of 8.0
Applications
• Phenol-Chloroform-Isoamyl alcohol mixture solution (25:24:1) is used in the extraction of nucleic acids, DNA and RNA
• When nucleic acids are extracted with the Phenol-Chloroform-Isoamyl alcohol mixture solution, a clear component separation is achieved. The proteins present in the sample are denatured and removed (collected in the organic phase or in the interphase), while nucleic acids remain in the aqueous phase. The chloroform denatures proteins and facilitates the separation of the aqueous and organic phases, and the isoamyl alcohol reduces foaming during extraction.
• It is also used to generate DNA minisatellite fingerprints
RUO – Research Use Only
Literature References
McKiernan, H.; Danielson, P. Chapter 21 - Molecular Diagnostic Applications in Forensic Science. Molecular Diagnostics (Third Edition). 2017, 371-394.
Zezza, F.; Pascale, M.; Mulè, G.; Visconti, A. Detection of Fusarium culmorum in wheat by a surface plasmon resonance-based DNA sensor. Journal of microbiological methods. 2006, 66(3), 529-537.