Camostat methanesulfate, 98%
Camostat methanesulfate, 98%
Camostat methanesulfate, 98%
Thermo Scientific Chemicals

Camostat methanesulfate, 98%

An orally active synthetic serine protease inhibitor | CAS: 59721-29-8 | C21H26N4O8S | 494.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62362.MC
also known as J62362-MC
Price (USD)
362.65
Special offer
Online exclusive
Ends: 30-Jun-2026
426.00
Save 63.35 (15%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
362.65
Special offer
Online exclusive
Ends: 30-Jun-2026
426.00
Save 63.35 (15%)
Each
Chemical Identifiers
CAS59721-29-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Infrared spectrumConforms
Assay from Supplier's CofA≥97.5% (HPLC)
It is used as a protease and Trypsin inhibitor. Furthermore it was found that this water induction and bicarbonate secretion increase was achieved via direct regulation of ductular cells of the dog pancreas. Other studies have shown Camostat mesylate to suppress spinal Fos expression in the rat, which is a marker for neuronal activation. In addition it was shown that this compound may prevent or depress pancreatitis-induced referred allodynia/hyperalgesia in rat models. Camostat mesylate has also demonstrated the ability to prevent Trypsin attack of microspheres, which causes the degradation product N-benzoyl-dl-arginine. Camostat mesylate is an inhibitor of KLK, plasminogen, Prothrombin and Trypsin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a protease and Trypsin inhibitor. Furthermore it was found that this water induction and bicarbonate secretion increase was achieved via direct regulation of ductular cells of the dog pancreas. Other studies have shown Camostat mesylate to suppress spinal Fos expression in the rat, which is a marker for neuronal activation. In addition it was shown that this compound may prevent or depress pancreatitis-induced referred allodynia/hyperalgesia in rat models. Camostat mesylate has also demonstrated the ability to prevent Trypsin attack of microspheres, which causes the degradation product N-benzoyl-dl-arginine. Camostat mesylate is an inhibitor of KLK, plasminogen, Prothrombin and Trypsin.

Solubility
Soluble in water (100 mM), DMSO (100 mM), and ethanol (<1mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Horiuchi et al. Gabexate and camostat, synthetic proteinase inhibitors, as direct inducing factors of water and bicarbonate secretion in the isolated and blood-perfused dog pancreas. J.Pharmacol.Exp.Ther. 1990, 252(1), 320-6.
  2. Ishikura et al. The proteinase inhibitor camostat mesilate suppresses pancreatic pain in rodents. Life Sci. 2007, 80(21), 1999-2004.